Learn More
CSF2, Rat, Clone: KT37, Abnova™
Rat monoclonal antibody raised against recombinant CSF2.
Brand: Abnova MAB9732.100ug
Description
The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes and macrophages. The active form of the protein is found extracellularly as a homodimer. This gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. [provided by RefSeq
Specifications
| CSF2 | |
| Monoclonal | |
| 1 mg/mL | |
| Rat monoclonal antibody raised against recombinant CSF2. | |
| In PBS (0.09% sodium azide) | |
| GMCSF/MGC131935/MGC138897 | |
| Rat | |
| Protein G purification | |
| RUO | |
| 1437 | |
| Store at 4°C for 1 month. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a |
| ELISA | |
| KT37 | |
| Unconjugated | |
| Sandwich ELISA (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CSF2 | |
| CSF2 | |
| Recombinant protein corresponding to human CSF2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.