missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gena (testicular germ cell-specific), Rat, Clone: TRA98, Abnova™
Rat monoclonal antibody raised against Gena (testicular germ cell-specific).
Brand: Abnova MAB7953.100ug
This item is not returnable.
View return policy
Specifications
| Gena (testicular germ cell-specific) | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rat | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| TRA98 | |
| Rat monoclonal antibody raised against Gena (testicular germ cell-specific). | |
| In PBS (50% glycerol) | |
| The hybridoma producing TRA98 was obtained by immunization of a rat with the cell lysate of adult mouse testis. | |
| RUO | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction