missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL2, Rat, Clone: JES6-1A12, Abnova™
Rat monoclonal antibody raised against recombinant Il2.
Brand: Abnova MAB3305.500ug
This item is not returnable.
View return policy
Specifications
| IL2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Il2 | |
| Il2 | |
| Recombinant protein corresponding to mouse Il2. | |
| RUO | |
| 16183 | |
| Mouse | |
| Liquid |
| Western Blot | |
| JES6-1A12 | |
| Rat monoclonal antibody raised against recombinant Il2. | |
| In 100mM BBS, pH 8.2 | |
| Il-2 | |
| Rat | |
| 500 μg | |
| Primary | |
| Mouse Interleukin-2 (IL-2). | |
| Store at 4°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction