missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL4, Rat, Biotin, Clone: BVD6-24G2, Abnova™
Rat monoclonal antibody raised against recombinant Il4.
Brand: Abnova MAB5877.500ug
This item is not returnable.
View return policy
Specifications
| IL4 | |
| Monoclonal | |
| Biotin | |
| The optimal working dilution should be determined by the end user. | |
| Il4 | |
| Il4 | |
| Recombinant protein corresponding to mouse Il4. | |
| RUO | |
| 16189 | |
| Mouse | |
| Liquid |
| ELISA | |
| BVD6-24G2 | |
| Rat monoclonal antibody raised against recombinant Il4. | |
| In PBS (0.09% sodium azide) | |
| Il-4 | |
| Rat | |
| 500 μg | |
| Primary | |
| Mouse Interleukin-4 (IL-4)1,2. This is a non-neutralizing antibody. | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction