Learn More
IL5, Rat, Clone: JES1-5A10, Abnova™
Rat monoclonal antibody raised against recombinant IL5.
Brand: Abnova MAB3294.500ug
Description
The protein encoded by this gene is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. This cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of this cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). This gene, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. This cytokine, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31. [provided by RefSeq
Specifications
| IL5 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| IL5 | |
| IL5 | |
| Recombinant protein corresponding to human IL5. | |
| RUO | |
| 3567 | |
| Human | |
| Liquid |
| ELISA | |
| JES1-5A10 | |
| Rat monoclonal antibody raised against recombinant IL5. | |
| In 100mM BBS, pH 8.2 | |
| EDF/IL-5/TRF | |
| Rat | |
| 500 μg | |
| Primary | |
| Human Interleukin-5. | |
| Store at 4°C. | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.