missing translation for 'onlineSavingsMsg'
Learn More

Itgam, Rat, APC-Cy7, Clone: M1/70, Abnova™

Product Code. 16291880
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16291880 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16291880 Supplier Abnova Supplier No. MAB9868.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat monoclonal antibody raised against Itgam.

Specifications

Antigen Itgam
Applications Flow Cytometry
Classification Monoclonal
Clone M1/70
Conjugate APC-Cyanine7
Description Rat monoclonal antibody raised against Itgam.
Dilution Flow Cytometry (≤ 0.3 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Itgam
Gene Alias CD11b/CD18/CR3/CR3A/Cd11b/F730045J24Rik/Ly-40/MAC1/Mac-1/Mac-1a
Gene Symbols Itgam
Host Species Rat
Immunogen Native Itgam from mouse splenic T cells and splenocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16409
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.