Learn More
Lpam-1, Rat, Biotin, Clone: DATK32, Abnova™
Rat monoclonal antibody raised against native Lpam-1.
Brand: Abnova MAB5796.500ug
This item is not returnable.
View return policy
Specifications
| Lpam-1 | |
| Monoclonal | |
| Biotin | |
| Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| CD49D | |
| Rat | |
| 500 μg | |
| Primary | |
| Mouse LPAM-1 (Integrin alpha4beta7). | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| DATK32 | |
| Rat monoclonal antibody raised against native Lpam-1. | |
| In PBS (0.09% sodium azide) | |
| Itga4 | |
| Native purified Lpam-1 from AKR/Cum mouse spontaneous T lymphoma cell line TK1. | |
| RUO | |
| 16401/16421 | |
| Mouse | |
| Liquid |