missing translation for 'onlineSavingsMsg'
Learn More

Mapre1, Rat, Clone: KT51, Abnova™

Product Code. 16160120
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16160120 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16160120 Supplier Abnova Supplier No. MAB1678.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat monoclonal antibody raised against recombinant Mapre1.

Specifications

Antigen Mapre1
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KT51
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Mapre1.
Dilution Western Blot (1:500-1:7000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Mapre1
Gene Alias 5530600P05Rik/AI462499/AI504412/AW260097/BIM1p/D2Ertd459e/Eb1
Gene Symbols Mapre1
Host Species Rat
Immunogen Recombinant GST fusion protein corresponding to amino acids 125-268 of mouse Mapre1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13589
Content And Storage Store at 4°C. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.