Learn More
CD45RB Rat anti-Mouse, BUV395, Clone: 16A, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 740211
Description
The 16A monoclonal antibody specifically recognizes an exon B-dependent epitope of CD45 lycoprotein, which is found at high density on peripheral B cells, T cytotoxic/suppressor cells, subset of T helper cells, and most thymocytes and at low density on macrophages and dendritic cells.The level of CD45RB expression appears to decrease as T lymphocytes progress from naive to memory cells. In addition, subpopulations of CD4+ T cells which express high and low levels of D45RB have different cytokine secretion profiles and mediate distinct immunological functions. CD25+ D4+ regulatory T (Treg) lymphocytes which control intestinal inflammation and autoimmunity express low levels of D45RB. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Specifications
| CD45RB | |
| Monoclonal | |
| 0.2mg/mL | |
| Ptprc; CD45R; CD45; LCA; Leukocyte common antigen; Ly-5; Lyt-4 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 16A | |
| Brilliant Ultraviolet 395 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Cloned Mouse TH2 cell lines | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only