Learn More
CD62P Rat anti-Mouse, Brilliant Violet 786, Clone: RB40.34, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 740884
Description
The RB40.34 monoclonal antibody specifically binds to mouse P-selectin (CD62P), a 140 kDa protein which is expressed on activated platelets, activated endothelial cells, and megakaryocytes. P-selectin mediates the adhesion of neutrophils and monocytes to activated platelets and endothelial cells, mediates leukocyte rolling, and is involved in the migration of leukocytes into inflamed tissues. CD24 and CD162 (PSGL-1) are ligands of CD62P. mAb RB40.34 can block mouse P-selectin binding to its ligands in vitro and in vivo.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
| CD62P | |
| Monoclonal | |
| 0.2mg/mL | |
| P-selectin; Selp; LECAM3; LYAM3; PADGEM; GMP-140; Grmp | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 λ |
| Flow Cytometry | |
| RB40.34 | |
| Brilliant Violet 786 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| P-selectin-IgG1 Fusion | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only