Learn More
CD86 Rat anti-Mouse, Brilliant Violet 786, Clone: PO3, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 740900
Description
The PO3 monoclonal antibody specifically binds to the B7-2 (CD86) costimulatory molecule, which is expressed on a broad spectrum of leukocytes, including B lymphocytes, T lymphocytes, macrophages, dendritic cells, and Langerhans cells, plus astrocytes. CD86 is expressed at low levels by freshly explanted peripheral B and T cells, and its expression is substantially increased by a variety of T and B cell-specific stimuli with a peak expression after 18-42 hours of culture. In contrast to most naive CD4+ T cells, memory CD4+ T cells express B7-2, both at the mRNA and protein levels. CD86, a ligand for CD28 and CTLA-4, is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. It has been shown to be involved in immunoglobulin class switching and triggering of mouse NK cell-mediated cytotoxicity. CD80 (B7-1) is an alternate ligand for CD28 and CTLA-4. PO3 antibody blocks the in vitro stimulation of T-cell proliferation of T-cell proliferation by soluble anti-CD3e antibody (mAb 145-2C11, Cat. No. 553057) in the presence of CD86-expressing accessory cells. In vivo administration of PO3 mAb can inhibit much of the autoantibody production in (NZB x NZW) F1 mice; and in combination with an anti-CD80 mAb, it can prevent the development and progression of mouse systemic lupus erythematosus-like autoimmune disease.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
| CD86 | |
| Monoclonal | |
| 0.2mg/mL | |
| Cd86; B7-2; B7.2; Ly58; Ly-58; ETC-1; MB7-2; TS/A2; Cd28l2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| PO3 | |
| Brilliant Violet 786 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| BALB/c Mouse B Leukemia Cell Line BCL1 | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only