missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgE Heavy Chain Secondary Antibody (LO-ME-3), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-68431R
This item is not returnable.
View return policy
Description
IgE Heavy Chain Monoclonal antibody specifically detects IgE Heavy Chain in Mouse samples. It is validated for ELISA, Western Blot (Negative)
Specifications
| IgE Heavy Chain | |
| Monoclonal | |
| DyLight 550 | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Purified IgE from BALB/c mice | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Western Blot (Negative) | |
| LO-ME-3 | |
| 50mM Sodium Borate | |
| Rat | |
| Affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction