missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgG Heavy Chain Secondary Antibody (LO-MG-7), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-42329
This item is not returnable.
View return policy
Description
IgG Heavy Chain Monoclonal antibody specifically detects IgG Heavy Chain in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications
| IgG Heavy Chain | |
| Monoclonal | |
| 1.0mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
| Rat | |
| IgM purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| LO-MG-7 | |
| Unconjugated | |
| Immunoglobulin G, ImmunoglobulinG | |
| Synthetic peptide encompassing the unique 20 N-terminal amino acid sequence of RTN1-C | |
| 0.2 mL | |
| Adaptive Immunity, Immunology | |
| Mouse Gamma Heavy Chain (gamma2a, gamma2b, gamma1 and gamma3) of Immunoglobulin (determined by immunodot). | |
| Store at 4C. Do not freeze. | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction