missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgG2a Heavy Chain Secondary Antibody (LO-MG2a-7), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-68435B
This item is not returnable.
View return policy
Description
IgG2a Heavy Chain Monoclonal antibody specifically detects IgG2a Heavy Chain in Mouse samples. It is validated for ELISA
Specifications
| IgG2a Heavy Chain | |
| Monoclonal | |
| Biotin | |
| Rat | |
| Affinity purified | |
| RUO | |
| Mouse | |
| Purified |
| ELISA | |
| LO-MG2a-7 | |
| PBS | |
| Purified IgG from BALB/c mice | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction