missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgM Heavy Chain Secondary Antibody (eB121-15F9), Janelia Fluor 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43282JF646
This item is not returnable.
View return policy
Description
IgM Heavy Chain Monoclonal antibody specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry
Specifications
| IgM Heavy Chain | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Mouse IgM | |
| 0.5 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| eB121-15F9 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction