Learn More
ORC3L, Rat, Clone: 1D6, Abnova™
Rat monoclonal antibody raised against ORC3L.
Brand: Abnova MAB2471.50uL
Description
The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Studies of a similar gene in Drosophila suggested a possible role of this protein in neuronal proliferation and olfactory memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene. [provided by RefSeq
Specifications
| ORC3L | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ORC3L | |
| ORC3L | |
| Human ORC3L. | |
| RUO | |
| 23595 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1D6 | |
| Rat monoclonal antibody raised against ORC3L. | |
| In PBS (0.09% sodium azide) | |
| LAT/LATHEO/ORC3 | |
| Rat | |
| 50 μL | |
| Primary | |
| human ORC3L. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.