Learn More
PROCR, Rat, Clone: LMR-42, Abnova™
Rat monoclonal antibody raised against full-length recombinant PROCR.
Brand: Abnova MAB3398.1mL
Description
The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy. [provided by RefSeq
Specifications
| PROCR | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Frozen sections) (1:20) The optimal working dilution should be determined by the end user. | |
| PROCR | |
| PROCR | |
| Recombinant protein corresponding to full length human PROCR. | |
| RUO | |
| 10544 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| LMR-42 | |
| Rat monoclonal antibody raised against full length recombinant PROCR. | |
| In serum-free culture supernatant (0.09% sodium azide) | |
| CCCA/CCD41/CD201/EPCR/MGC23024/bA42O4.2 | |
| Rat | |
| 1 mL | |
| Primary | |
| The rat monoclonal antibody LMR-42 detects an outer-surface epitope of a 55 KDa plasma membrane protein over-expressed in several human non-Pgp multidrug-resistant (MDR) tumor cell lines. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.