missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ptprc, Rat, Biotin, Clone: I3/2.3, Abnova™
Rat monoclonal antibody raised against native Ptprc.
Brand: Abnova MAB5937.500ug
This item is not returnable.
View return policy
Specifications
| Ptprc | |
| Monoclonal | |
| Biotin | |
| Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Ptprc | |
| Ptprc | |
| Native purified Ptprc from T1M1 (Thy-1-C) cells. | |
| RUO | |
| 19264 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| I3/2.3 | |
| Rat monoclonal antibody raised against native Ptprc. | |
| In PBS (0.09% sodium azide) | |
| B220/CD45R/Cd45/Ly-5/Lyt-4/T200/loc | |
| Rat | |
| 500 μg | |
| Primary | |
| Specificity Mouse CD45 (T200 or leukocyte common antigen/LCA, all isoforms) | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction