Learn More
Rat anti-Rabbit IgG (H+L) Secondary Antibody, Allophycocyanin/Cy7 (Pre-adsorbed), Novus Biologicals™
Rat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72765APCCY7
Product Code. 18580274
  • € 822.00 / 500µL
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
PBS
Rabbit IgG whole molecule
500 μg
Adaptive Immunity, Immunology
Rabbit
IgG
Western Blot, ELISA, Immunohistochemistry
APC-Cyanine7
Rat
Multi-step
RUO
Secondary
Store at 4°C in the dark. Do not freeze.