Learn More
TNFSF13, Rat, Clone: Sacha-1, Abnova™
Rat monoclonal antibody raised against partial recombinant TNFSF13.
Brand: Abnova MAB3364.100ug
Description
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) ligand family. This protein is a ligand for TNFRSF17/BCMA, a member of the TNF receptor family. This protein and its receptor are both found to be important for B cell development. In vitro experiments suggested that this protein may be able to induce apoptosis through its interaction with other TNF receptor family proteins such as TNFRSF6/FAS and TNFRSF14/HVEM. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. Other transcripts that skip the last exon of the upstream gene (TNFSF12) and continue with the second exon of this gene have been identified; such read-through transcripts are contained in GeneID 407977, TNFSF12-TNFSF13. [provided by RefSeq
Specifications
| TNFSF13 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TNFSF13 | |
| TNFSF13 | |
| Recombinant protein corresponding to chimeric protein with amino acids 105-250 of human TNFSF13l and amino acids 16-108 of human ACRP30 headless. | |
| RUO | |
| 8741 | |
| Human, Mouse | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| Sacha-1 | |
| Rat monoclonal antibody raised against partial recombinant TNFSF13. | |
| In PBS (0.02% sodium azide) | |
| APRIL/CD256/TALL2/TRDL-1/UNQ383/PRO715/ligand | |
| Rat | |
| 100 μg | |
| Primary | |
| Recognizes human and mouse APRIL. | |
| Store at 4°C. | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.