missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat beta 2-Microglobulin Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3864S-100UG
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Rat samples. It is validated for Flow Cytometry.
Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P07151 | |
| B2M | |
| Mouse myeloma cell line NS0-derived recombinant rat beta 2-Microglobulin Ile21-Met119 Accession # P07151 | |
| RUO | |
| 567 | |
| Rat | |
| IgM |
| Flow Cytometry | |
| 747502 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B2M, beta 2Microglobulin, beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects rat beta 2-Microglobulin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat alpha 2-Macroglobulin is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction