missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat C-Reactive Protein/CRP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1744-SP
This item is not returnable.
View return policy
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Rat samples. It is validated for Western Blot.
Specifications
| C-Reactive Protein/CRP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P48199 | |
| CRP | |
| Mouse myeloma cell line NS0-derived recombinant rat CRP His20-Ser230 Accession # P48199 | |
| 25 μg | |
| Primary | |
| Detects rat CRP in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human CRP, recombinant mouse CRP, or recombinant porcine CRP. | |
| Rat | |
| Purified |
| Western Blot | |
| 233502 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1401 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction