missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CCL3/MIP-1 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB66251-SP
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Rat samples. It is validated for Immunohistochemistry.
Specifications
| CCL3/MIP-1 alpha | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P50229 | |
| CCL3 | |
| E. coli-derived recombinant rat CCL3/MIP-1 alpha Ala24-Ala92 Accession # P50229 | |
| 25 μg | |
| Primary | |
| Detects rat CCL3/MIP-1 alpha in direct ELISAs. In direct ELISAs, less than 50% cross-reactivity with recombinant mouse and cotton rat CCL3/MIP-1 alpha is observed, and no cross-reactivity with recombinant human, canine, and HHV CCL3/MIP-1 alpha, recombinant rat CCL4/MIP-1 beta, or recombinant mouse CCL5/RANTES is observed. | |
| Rat | |
| Purified |
| Immunohistochemistry | |
| 769303 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6348 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction