missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat CXCL1 (GRO alpha) Recombinant Protein

Product Code. 10641884
Click to view available options
Quantity:
25 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10641884

Brand: Gibco™ PRC1066

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Recombinant rat GRO should be reconstituted in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Recombinant rat GRO is also soluble in most buffered solutions. Further dilutions should be made in buffered solution with carrier protein such as PBS with 1% BSA. Storage: This lyophilized preparation is stable at 2°C to 8°C, but should be kept at -20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at -20°C. Avoid repeated freeze/thaw cycles.

CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14095
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 81503
Molecular Weight (g/mol) 7.8 kDa
Name Rat CXCL1 (GRO alpha)
Purification Method Purified
Quantity 25 μg
Storage Requirements 4°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO α; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROα; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51
Common Name CXCL1 (GRO alpha)
Gene Symbol Cxcl1
Biological Activity ED50 = 10 - 100 ng/mL; determined by the dose-dependent chemotaxis of rat neutrophils.
Product Type Protein
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Sequence APVANELRCQ CLQTVAGIHF KNIQSLKVMP PGPHCTQTEV IATLKNGREA CLDPEAPMVQ KIVQKMLKGV PK
Content And Storage 4°C
Classification Carrier-Free
Activity ED50 = 10 - 100 ng/mL; determined by the dose-dependent chemotaxis of rat neutrophils.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Chemokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype GRO⁄KC⁄CINC Chemokines
Research Category Cell Cycle & Proliferation, Oncology, Inflammation, Stem Cell Research, Neurobiology, Angiogenesis
Product Line Gibco
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.