missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CXCR2/IL-8RB Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8110RN-100UG
This item is not returnable.
View return policy
Description
CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Rat samples. It is validated for Flow Cytometry.
Specifications
| CXCR2/IL-8RB | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, CMKAR2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 RB, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
| NS0 mouse myeloma cell line transfected with rat CXCR2/IL-8 RB Accession # P35407 | |
| 100 μg | |
| 3579 | |
| Rat | |
| Supernatant |
| Flow Cytometry | |
| 866614R | |
| Flow Cytometry 0.25-1μg/106 cells | |
| P35407 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects rat CXCR2/IL-8 RB in direct ELISAs. In Flow Cytometry, stains HEK293 cells transfected with rat CXCR2/IL-8 RB, but does not stain non-transfected cells. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction