missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CXCR6 Alexa Fluor 594-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB8196RT-100UG
This item is not returnable.
View return policy
Description
CXCR6 Monoclonal specifically detects CXCR6 in Rat samples. It is validated for Flow Cytometry.
Specifications
| CXCR6 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human CXCR6 in ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 879112R | |
| Flow Cytometry 0.25-1μg/106 cells | |
| Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
| NS0 mouse myeloma cell line transfected with rat CXCR6 | |
| 100 μg | |
| 10663 | |
| Rat | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction