missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CXCR7/RDC-1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems FAB8399N-100UG
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Rat samples. It is validated for Flow Cytometry.
Specifica
| CXCR7/RDC-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_445804 | |
| CXCR7 | |
| NS0 mouse myeloma cell line transfected with rat CXCR7/RDC-1 Met1-Lys362 Accession # NP_445804 | |
| RUO | |
| 57007 | |
| Rat | |
| IgG2a |
| Flow Cytometry | |
| 896032 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains rat CXCR7/RDC-1 transfectants but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only (RUO)
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto