missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Rat Fgf2 (P13109, 10 a.a. - 154 a.a.) Partial Recombinant Protein

Product Code. 16222190
Click to view available options
:
50μg
This item is not returnable. View return policy

Product Code. 16222190

Brand: Abnova™ P5037.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Sequence: PALPEDGGGAFPPGHFKDPKRLYCKNGGFFLRIHPDGRVDGVREKSDPHVKLQLQAEERGVVSIKGVCANRYLAMKEDGRLLASKCVTEECFFFERLESNNYNTYRSRKYSSWYVALKRTGQYKLGSKTGPGQKAILFLPMSAKS

Specifications

Accession Number P13109
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 54250
Molecular Weight (g/mol) 16kDa
Name Fgf2 (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 50 ug
Immunogen PALPEDGGGAFPPGHFKDPKRLYCKNGGFFLRIHPDGRVDGVREKSDPHVKLQLQAEERGVVSIKGVCANRYLAMKEDGRLLASKCVTEECFFFERLESNNYNTYRSRKYSSWYVALKRTGQYKLGSKTGPGQKAILFLPMSAKS
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration Endotoxin content was assayed using a LAL gel clot method. Endotoxin level was found to be <0.1ng/μg (1EU/μg).
Gene Alias Fgf-2/bFGF
Common Name Fgf2
Gene Symbol Fgf2
Biological Activity The ED(50) determined by dose-dependent stimulation of thymidine uptake by 3T3 cells expressing FGF receptors, was ofund to be < 1ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.