missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat GM-CSF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB5181
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Rat samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| GM-CSF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1437 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 83308 | |
| Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P48750 | |
| CSF2 | |
| E. coli-derived recombinant rat GM-CSF Ala1-Lys127 Accession # P48750 | |
| 500 μg | |
| Primary | |
| Detects rat GM-CSF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant GM-CSF from mouse, human, or pig is observed. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction