missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Growth Hormone R/GHR Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1211
This item is not returnable.
View return policy
Description
Growth Hormone R Polyclonal specifically detects Growth Hormone R in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications
| Growth Hormone R | |
| Polyclonal | |
| Unconjugated | |
| P16310 | |
| GHR | |
| Mouse myeloma cell line NS0-derived recombinant rat Growth Hormone R/GHR Phe19-Arg265 Accession # P16310 | |
| 100 μg | |
| Primary | |
| Detects rat Growth Hormone R/GHR in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse GHR and recombinant human GHR is observed. | |
| Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.075-0.3 ug/mL | |
| GH receptor, GHBP, GHR, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2690 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction