Learn More
Invitrogen™ IgA Rat Uncoated ELISA Kit with Plates
For quantitative detection of rat IgA.
€ 240.00 - € 782.00
Specifications
Assay Sensitivity | 16ng/mL |
---|---|
Sample Type | Cell Culture Supernatant, Plasma, Serum |
Target Species | Rat |
For Use With (Application) | ELISA |
Storage Requirements | 2°C to 8°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15510917
|
Invitrogen™
88-50480-22 |
2 x 96 Tests |
€ 240.00
Pack of 2 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15540917
|
Invitrogen™
88-50480-86 |
10 x 96 Tests |
€ 544.00
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15520917
|
Invitrogen™
88-50480-76 |
20 x 96 Tests |
€ 782.00
Pack of 20 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
The Rat Total Immunoglobulin G (IgG) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat total IgG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rat IgA Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
16ng/mL | |
Rat | |
2°C to 8°C | |
IgA |
Cell Culture Supernatant, Plasma, Serum | |
ELISA | |
Immunoglobulin A |
For Research Use Only