missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IgE ELISA Kit

Product Code. 18057404
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18057404 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18057404 Supplier Invitrogen™ Supplier No. EEL126

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Rat IgE, also referred to as rat immunoglobulin E, is an antibody isotype synthesized by rats in response to allergens or parasites. It plays a pivotal role in mediating allergic reactions and immune responses against specific pathogens. IgE antibodies are primarily responsible for eliciting the release of inflammatory mediators, such as histamine, from mast cells and basophils. This immune response can give rise to symptoms like pruritus, edema, and bronchoconstriction. Investigating the genetic target Rat IgE can provide valuable insights into the underlying mechanisms of allergic diseases and contribute to the development of innovative therapeutic strategies for allergies and immunological disorders in both rats and humans.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01855
Assay Range 1.56 to 100 ng/mL
Assay Sensitivity 0.94 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene Symbol IGHE
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Rat IgE
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.