missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), APC, eBioscience™, Invitrogen™
Rat Isotype Control
€ 152.00 - € 252.00
Specifications
Antigen | Rat IgG2b kappa |
---|---|
Clone | eB149/10H5 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | APC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15330720
|
Affymetrix eBioscience
17-4031-81 |
50 μg |
€ 152.00
50µg |
In Stock
Log in to see stock. |
|||||
15340720
|
Affymetrix eBioscience
17-4031-82 |
100 μg |
€ 252.00
100µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: APC Rat IgG2b Isotype Control has been reported for use in flow cytometric analysis. Applications Tested: Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.Specifications
Rat IgG2b kappa | |
0.2 mg/mL | |
APC | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eB149/10H5 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.