missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-1 alpha/IL-1F1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF500
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Polyclonal specifically detects IL-1 alpha/IL-1F1 in Rat samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-1 alpha/IL-1F1 | |
| Polyclonal | |
| Unconjugated | |
| P16598 | |
| IL1A | |
| E. coli-derived recombinant rat IL-1 alpha/IL-1F1 Ser115-Ser270 Accession # P16598 | |
| 100 μg | |
| Primary | |
| Detects rat IL-1 alpha/IL-1F1 in direct ELISAs and Western blots. In direct ELISAs, less than 50% cross-reactivity with recombinant mouse IL-1 alpha/IL-1F1 and recombinant cotton rat IL-1 alpha/IL-1F1 is observed, and less than 15% cross-reactivity with recombinant human IL-1 alpha/IL-1F1 and recombinant porcine IL-1 alpha/IL-1F1 is observed. | |
| Rat | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.003-0.009 ug/mL | |
| BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3552 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction