missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-12 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1760-SP
This item is not returnable.
View return policy
Description
IL-12 Polyclonal specifically detects IL-12 in Rat samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-12 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3592 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.05-0.25 ug/mL | |
| p40: NP_072133; p35: Q9R103 | |
| IL12A | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat IL-12 p40: Met23-Ser335; p35: Arg23-Ser215 Accession # p40: NP_072133; p35: Q9R103 | |
| 25 μg | |
| Primary | |
| Detects rat IL-12 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse IL-12 and less than 5% cross-reactivity with recombinant human IL-12 is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction