missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat IL-2, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17896538
Click to view available options
Quantity:
250 μg
5 μg
Unit Size:
250µg
5µg
This item is not returnable. View return policy

Product Code. 17896538

Brand: Gibco™ AF40002250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P17108
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 116562
Molecular Weight (g/mol) 15.3 kDa
Name Rat IL-2, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 250 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Common Name IL-2
Gene Symbol IL2
Biological Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAIAQSI ISTM
Content And Storage -20°C
Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 10^6 units/mg
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.