Learn More
Rat IL-7R alpha/CD127 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB8484P-100
 View more versions of this product
Product Code. 18115011
  • € 507.00 / 100 tests
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-7R alpha/CD127
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3575
Rat
Purified
Flow Cytometry
717519
Flow Cytometry 10 uL/10^6 cells
NP_001099888
IL7R
Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127 Glu21-Asp239 Accession No. NP_001099888
100 Tests
Primary
Detects rat IL-7 R alpha/CD127 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a