missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat LIX Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF543
This item is not returnable.
View return policy
Description
LIX Polyclonal specifically detects LIX in Rat samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| LIX | |
| Polyclonal | |
| Biotin | |
| P97885 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6374 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| Cxcl6 | |
| E. coli-derived recombinant rat LIX Ala38-Arg125 Accession # P97885 | |
| 50 μg | |
| Primary | |
| Detects rat LIX in ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse CXCL5/GCP2 is observed, and less than 5% cross-reactivity with recombinant human CXCL6/GCP2 is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction