missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems Rat Luminex™ Discovery Assay
Specifications
Specifications
| Quantity | 9 Plex |
| Species | Rat |
| Binding Target | Multiplex Cytokine Assay Kits |
| Content And Storage | Store the unopened kit at 2-8 °C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?