missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat Luteinizing Hormone ELISA Kit (Chemiluminescence)

Product Code. 18690351 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18690351

Brand: Novus Biologicals™ NBP268054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Chemiluminescence)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.31 to 20 mIU/mL
Assay Sensitivity 0.19 mIU/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Rat LH in samples. No significant cross-reactivity or interference between Rat LH and analogues was observed. Typical data
For Use With (Application) ELISA
Gene ID (Entrez) 3972
Quantity 1 Kit
Research Discipline Cancer, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CGB4;HH23;LHB;LSH-B;LSH-beta;Lutropin subunit beta
Target Rat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.