missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Neurexin 1 beta/NXRN1b Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4524-SP
This item is not returnable.
View return policy
Description
Neurexin 1 beta/NXRN1b Polyclonal specifically detects Neurexin 1 beta/NXRN1b in Rat samples. It is validated for Western Blot.
Specifications
| Neurexin 1 beta/NXRN1b | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Hs.22998, NXR1B, NXRN1b, PTHSL2, SCZD17 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9378 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q63373 | |
| Nrxn1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neurexin 1 beta/NXRN1b Ala47-Thr392 Accession # Q63373 | |
| 25 μg | |
| Primary | |
| Detects rat Neurexin 1 beta/NXRN1b in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant rat Neurexin 1 alpha is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction