missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Neuropilin-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB566
This item is not returnable.
View return policy
Description
Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Rat samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifications
| Neuropilin-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8829 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 130603 | |
| Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction 5 ug/mL, CyTOF-ready | |
| Q9QWJ9 | |
| NRP1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-1 Phe22-Asp854 (Lys811Arg, Pro812-Gly828 del), predicted Accession # Q9QWJ9 | |
| 500 μg | |
| Primary | |
| Detects rat Neuropilin-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat Neuropilin-2 is observed. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction