missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat Osteocalcin ELISA Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
Description
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
Specifications
Specifications
| Accession Number | P04640 |
| Assay Range | 0.78 to 50 ng/mL |
| Assay Sensitivity | 0.47 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 25295 |
| Gene Symbol | BGLAP |
| Interassay CV | <10% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction