missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Serpin A6 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3838-SP
This item is not returnable.
View return policy
Description
Serpin A6/Cortisol Binding Globulin Polyclonal specifically detects Serpin A6/Cortisol Binding Globulin in Rat samples. It is validated for Western Blot.
Specifications
| Serpin A6/Cortisol Binding Globulin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antitrypsin), member 6, CBG, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 866 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_001009663 | |
| SERPINA6 | |
| Mouse myeloma cell line NS0-derived recombinant rat Serpin A6 Ser23-Ala396 Accession # NP_001009663 | |
| 25 μg | |
| Primary | |
| Detects rat Serpin A6 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) Serpin A1, rmSerpin A5, and recombinant human Serpin A4 is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction