missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat TIM-1/KIM-1/HAVCR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Brand: R&D Systems AF3689-SP
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Polyclonal specifically detects TIM-1/KIM-1/HAVCR in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TIM-1/KIM-1/HAVCR | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 26762 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| O54947 | |
| HAVCR1 | |
| Mouse myeloma cell line NS0-derived recombinant rat TIM-1/KIM-1/HAVCR Ser18-Val238 Accession # O54947 | |
| 25 μg | |
| Primary | |
| Detects rat TIM-1/KIM-1/HAVCR in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) TIM-1 is observed, and less than 1% cross-reactivity with recombinant human TIM-1, recombinant rat TIM-2, rmTIM-3, rmTIM-4, rmTIM-6, and rmTIM-7 is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction