missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat Total MMP-9 Quantikine ELISA Kit

Product Code. 18771994
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18771994

Brand: R&D Systems™ RMP900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Total MMP-9 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat Total MMP-9 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat MMP-9 in cell culture supernates (50 ul), serum (10 ul), platelet-poor heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.028 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Platelet-poor Plasma (Heparin), Serum
Specificity Natural and Recombinant Rat MMP-9 (Pro-, active, and TIMP-complexed)
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 4318
Gene Symbol Mmp9
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),Platelet-poor Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase
Target MMP-9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.