Learn More
Gibco™ Rat (Wistar) Cryopreserved Hepatocytes, Plateable Female
Click to view available options
Unit Size:
1mL
Description
These plateable cryopreserved female rat (Wistar) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.
Cells per vial: 4–8 million
Get the Results You Need with GIBCO™ Hepatocytes
High quality hepatocytes with high viabilities, in vivo-like enzymee xpression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure that you receive the highest quality cryopreserved hepatocytes.
Each lot is tested for:
Phase I and II enzyme activities
Proper morphology
Viabilities generally >75%
Attachment efficiency
Monolayer formation and integrity
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Links:
Learn more about our cryopreserved hepatocytes.
Learn about our full line of hepatocytes.
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
Cells per vial: 4–8 million
Get the Results You Need with GIBCO™ Hepatocytes
High quality hepatocytes with high viabilities, in vivo-like enzymee xpression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure that you receive the highest quality cryopreserved hepatocytes.
Each lot is tested for:
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Links:
Learn more about our cryopreserved hepatocytes.
Learn about our full line of hepatocytes.
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
Specifications
Specifications
| Content And Storage | Cryopreserved Hepatocytes stored in liquid nitrogen (LN2). |
| Cell Type | Hepatocytes |
| Form | Cryopreserved |
| Species | Rat |
| Donor Source | Pooled |
| Product Line | Gibco |