Learn More
Rauwolscine hydrochloride, 99%, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar J63198.MC
Quantity 100mg
Product Code. 15475459
  • ONLINE EXCLUSIVE PRICE
    € 138.00
    € 135.00 / 100mg
    Save € 3.00
    2% Off
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6211-32-1
MFCD06668052
UN1544
17?-hydroxy-20?-yohimban-16?-carboxylic acid,methyl ester hydrochloride
COC(=O)C1C(CCC2C1CC3C4=C(CCN3C2)C5=CC=CC=C5N4)O.Cl
390.908
390.91
Rauwolscine hydrochloride
C21H27ClN2O3
100mg
Light sensitive
PIPZGJSEDRMUAW-XQDOVXNESA-N
methyl (1S,15S,18S,19S)-18-hydroxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate;hydrochloride
90479742
99%
Product Suggestions
Product Suggestions
Videos
SDS Documents