missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAVER1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22291AF532
This item is not returnable.
View return policy
Description
RAVER1 Polyclonal antibody specifically detects RAVER1 in Human samples. It is validated for Western Blot
Specifications
| RAVER1 | |
| Polyclonal | |
| Western Blot | |
| KIAA1978Protein raver-1, ribonucleoprotein PTB-binding 1, ribonucleoprotein, PTB-binding 1 | |
| The immunogen this antibody was made to, maps to a region between residue 556 and 606 of human Ribonucleoprotein, PTB-Binding using the numbering given in entry Q8IY67.1 (GeneID 125950). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 125950 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction