missing translation for 'onlineSavingsMsg'
Learn More

RAVER1 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18634062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18634062 0.02 mL 0.02mL
18664792 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18634062 Supplier Novus Biologicals Supplier No. NBP2944130.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAVER1 Polyclonal antibody specifically detects RAVER1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAVER1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias KIAA1978Protein raver-1, ribonucleoprotein PTB-binding 1, ribonucleoprotein, PTB-binding 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 390-490 of human RAVER1 (NP_597709.2). GPALSTALLQLALQTQGQKKPGILGDSPLGALQPGAQPANPLLGELPAGGGLPPELPPRRGKPPPLLPSVLGPAGGDREALGLGPPAAQLTPPPAPVGLRG
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125950
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.